Merck
  • Role of endogenous cyclo(His-Pro) in cold-induced hypothermia in the desert rat (Mastomys natalensis).
Role of endogenous cyclo(His-Pro) in cold-induced hypothermia in the desert rat (Mastomys natalensis).
Peptides (1994-01-01)
R Shukla, N H Rahmani, H Mizuma, R C Srimal, C Prasad
ABSTRACT
Central administration of exogenous cyclo(His-Pro) (CHP) is known to produce hypothermia in rodents. In the present study, we examined the role of endogenous CHP in cold-induced hypothermia in the desert rat, Mastomys natalensis. The results of these studies show that a rise in hypothalamic CHP content accompanied a decrease in rectal temperature during cold exposure. Immunoneutralization of endogenous CHP resulted in a significant decline in cold-induced hypothermia. In addition, central administration of cyclo(Ala-Gly), a structural analogue of CHP, also led to a decrease in cold-induced hypothermia. The results of these studies show that changes in endogenous CHP levels may affect body temperature regulation.
Sigma-Aldrich
Glycine anhydride, cyclic dipeptide
Pricing and availability is not currently available.