Merck
  • Analysis of mast cell subpopulations (MCT, MCTC) in cutaneous inflammation using novel enzyme-histochemical staining techniques.
Analysis of mast cell subpopulations (MCT, MCTC) in cutaneous inflammation using novel enzyme-histochemical staining techniques.
Experimental dermatology (1994-12-01)
B Algermissen, F Bauer, D Schadendorf, J D Kropp, B M Czarnetzki
ABSTRACT
In order to gain insights into the dynamics of mast cell subpopulations in normal and diseased skin, a novel enzyme-histochemical double and triple staining method was employed that allowed the detection of metachromasia (toluidine blue) and the mast cell proteases tryptase and chymase within the same cell. Cryostat sections were used of skin biopsies from the following specimens: normal skin (N = 4), psoriasis (N = 13), atopic eczema (N = 7), lichen planus (N = 6), interferon alpha 2a injection sites (N = 1) of a leukemic infiltrate and corresponding normal skin of the same patient before and after treatment. (i) Equal numbers of tryptase- and chymase-positive mast cells (MCTC) were obtained in all normal and diseased specimens in papillary and reticular dermis, with threefold increases around appendages. (ii) Tryptase-positive mast cells (MCT) were absent in normal skin, but were markedly increased in a disease-specific pattern within the papillary dermis, the inflammatory infiltrate and around appendages. (iii) Marked increases of MCT were also noted at interferon injection sites within the leukemic infiltrate, but not in the normal skin of the same patient. These data suggest that disease-dependent mast cell dynamics involve only MCT in cutaneous inflammation and that MCT numbers are controlled by distinct, disease-specific local tissue factors.
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, Dye content ≥80 %
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, for microscopy (Hist.)