Merck
  • Accumulation of a commercial polychlorinated biphenyl mixture (Aroclor 1016) in adult rhesus monkeys and their nursing infants.
Accumulation of a commercial polychlorinated biphenyl mixture (Aroclor 1016) in adult rhesus monkeys and their nursing infants.
Toxicology (1984-02-14)
D A Barsotti, J P Van Miller
ABSTRACT
Female rhesus monkeys (24) were divided into 3 groups and fed diets calculated to contain 1.0, 0.25 or 0 parts per million (ppm) of a commercial mixture of polychlorinated biphenyls (PCBs), Aroclor 1016. The animals consumed a calculated total of 18.1 +/- 3.1, 4.5 +/- 0.6 and 0 mg Aroclor 1016/kg of body weight over the 87 +/- 9 week experimental period. All animals were bred, conceived and experienced uncomplicated pregnancies. The birth weights of the infants born to females receiving the 1.0 ppm PCB diets were significantly less than those of the control infants. There was a positive relationship between the levels of Aroclor 1016 in the tissues and the dose administered in the diets. Aroclor 1016 content of infant tissues was consistent with maternal adipose tissue PCB levels. The content of Aroclor 1016 in the milk fat of the animals more closely approximated that in the individual's adipose tissue than in the serum. At weaning, the mesenteric fat of the experimental infants contained concentrations of Aroclor 1016 which were 4-7 times that found in the mothers. Analyses of the tissues and fluids of the experimental animals demonstrated accumulation of PCB isomerides. The number of isomerides present and the ratio of those detected differed between adult and infants as well as between milk and serum. After weaning, the pattern of Aroclor 1016 isomerides present in the infants' adipose was similar to that observed at weaning although the concentrations had decreased.
Product Description
Supelco
Aroclor 1016 solution, certified reference material, 1000 μg/mL in isooctane
Supelco
Aroclor 1016 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1016 solution, certified reference material, 200 μg/mL in methanol