Merck
  • Adverse effects of artificial buffers on contractile responses of arterial and venous smooth muscle.
Adverse effects of artificial buffers on contractile responses of arterial and venous smooth muscle.
British journal of pharmacology (1980-06-01)
B M Altura, B T Altura, A Carella, P D Turlapaty
ABSTRACT
1 In vitro studies were undertaken on rat aortic strips and portal vein segments in order to determine whether or not several commonly used artificial buffers, i.e., tris(hydroxymethyl) aminomethane (Tris), N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid (HEPES), morpholine propanesulphonic acid (MOPS), N,N bis(2-hydroxyethyl) glycine (BICINE) and 1,4-piperazinediethanesulphonic acid (PIPES), can exert direct actions on vascular smooth muscle. 2 All artificial buffers used in 5 mM concentrations were found to inhibit development of spontaneous mechanical activity. 3 Tris, HEPES, MOPS, BICINE and PIPES markedly attenuated contractions induced by adrenaline, angiotensin and KCl. The fast phase components of the agonist-induced contractions were either obliterated or reduced in the presence of the artificial buffers. The sustained slow phase components were greatly reduced and retarded by the artificial buffers. 4 The relative order of artificial buffer potency (i.e., from 100% to 14% inhibition) seems to depend upon the agonist and type of smooth muscle. 5 All of these inhibitory effects were reversible, since normal contractile responses and spontaneous mechanical activity could be obtained by simply reincubating the smooth muscles in Krebs-Ringer bicarbonate buffer. 6 A variety of pharmacological antagonists failed to mimic or affect the inhibitory effects of Tris, HEPES, MOPS, PIPES and BICINE. 7 These data show that five of the most commonly used artificial buffers, to study muscles in vitro, exert adverse effects on contractility of arterial and venous smooth muscle.
Sigma-Aldrich
BICINE, BioUltra, ≥99.5% (T)
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)
Sigma-Aldrich
BICINE, ≥99% (titration)