Merck
  • Hepatic microsomal O-de-ethylases in cod (Gadus morhua): their induction by Aroclor 1254 but not by Aroclor 1016.
Hepatic microsomal O-de-ethylases in cod (Gadus morhua): their induction by Aroclor 1254 but not by Aroclor 1016.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology (1983-01-01)
P D Hansen, R F Addison, D E Willis
ABSTRACT
1. The characteristics of hepatic ethoxycoumarin and ethoxyresorufin O-de-ethylases in juvenile cod (Gadus morhua) from the North Sea are described. 2. Feeding Aroclor 1254 to juvenile cod to produce liver concentrations of approx 900 microgram X g-1 (wet wt) induced ethoxycoumarin O-de-ethylase approx 30-fold, but had no effect on ethoxyresorufin O-deethylase activity. 3. Feeding Aroclor 1016 to juvenile cod to produce liver concentrations of approx 300 micrograms X g-1 (wet wt.) did not induce ethoxycoumarin O-de-ethylase activity.
Product Description
Supelco
Aroclor 1016 solution, certified reference material, 1000 μg/mL in isooctane
Supelco
Aroclor 1016 solution, certified reference material, 200 μg/mL in methanol
Supelco
Aroclor 1016 solution, certified reference material, 50 mg/kg in transformer oil