Merck
  • Increased safety and specificity in the thin-layer chromatographic identification of marihuana.
Increased safety and specificity in the thin-layer chromatographic identification of marihuana.
Journal of forensic sciences (1979-10-01)
R B Hughes, M S Kessler, R R Kessler
PMID541645
ABSTRACT
We are reporting what we believe to be a safe, rapid TLC system and spray for the identification of marihuana. The use of Fast Blue 2B salt greatly enhances the specificity of the TLC analysis of suspected marihuana samples.
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, Dye content ≥80 %
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt