Merck
  • Deamination of aliphatic amines by monoamine oxidase A and B studied using a bioluminescence technique.
Deamination of aliphatic amines by monoamine oxidase A and B studied using a bioluminescence technique.
Journal of neurochemistry (1985-05-01)
M Tenne, M B Youdim, S Ulitzur, J P Finberg
ABSTRACT
Deamination of n-octylamine and n-decylamine has been studied in various tissues using a new bioluminescence technique. Selectivity of n-octylamine and n-decylamine as substrates for monoamine oxidase (MAO) A or B has been determined using both clorgyline and (-)-deprenyl inhibition curves and kinetic parameters. Homogenates of rat brain, liver and heart containing predominantly MAO-A or -B were prepared by preincubation for 60 min with (-)-deprenyl or clorgyline (30 nM), respectively. Human placenta (MAO-A) and platelet (MAO-B) were used as reference tissues containing only one MAO form. In tissues (rat liver, brain) containing both MAO forms in equal proportion, inhibition curve studies showed a preference of both substrates for the B form of the enzyme; however, where MAO-A was the major form (rat heart, human placenta), clorgyline was the more effective inhibitor. In the beef brain cortex n-octylamine showed marked preference for MAO-B, whereas n-decylamine was selective toward-MAO-A. Kinetic studies in general supported the picture of greater selectivity of the aliphatic amine substrates for deamination by MAO-B, as reflected by lower Km values for this enzyme type. However, n-octylamine was more selective for MAO-B than n-decylamine in both kinetic and inhibition curve studies. The deamination of these aliphatic amine substrates cannot be explained only by reference to the binary classification of MAO into types A and B.
Sigma-Aldrich
Decylamine, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Decylamine, 95%
Pricing and availability is not currently available.