Merck
  • Prevalence of occupational asthma in spray painters exposed to several types of isocyanates, including polymethylene polyphenylisocyanate.
Prevalence of occupational asthma in spray painters exposed to several types of isocyanates, including polymethylene polyphenylisocyanate.
Journal of occupational medicine. : official publication of the Industrial Medical Association (1987-04-01)
P Séguin, A Allard, A Cartier, J L Malo
ABSTRACT
The prevalence of occupational asthma was assessed in four paint shops of a large assembly plant where 51 employees were exposed to several types of isocyanates, including polymethylene polyphenylisocyanate (PPI). Three employees were first referred by their physician for asthma symptoms. A questionnaire was administered to the other 48 employees. Seven of these were suspected of having work-related asthma. Airway hyperexcitability to inhaled histamine was demonstrated in these ten subjects (three referred and seven screened). The diagnosis of occupational asthma was confirmed in six subjects (three referred and three screened) through specific inhalation challenges in the laboratory to a paint system component containing PPI. Thus, the prevalence of occupational asthma was 11.8% in these paint shops using several types of isocyanates, including PPI.
Sigma-Aldrich
Poly[(phenyl isocyanate)-co-formaldehyde], average Mn ~400