- Evaluation of the Fourier transform infrared (FTIR) spectrophotometer for analysis of trichloroethylene (TCE) in the presence of Freon-113 in carbon disulfide eluates of charcoal air sampling tubes.
Evaluation of the Fourier transform infrared (FTIR) spectrophotometer for analysis of trichloroethylene (TCE) in the presence of Freon-113 in carbon disulfide eluates of charcoal air sampling tubes.
American Industrial Hygiene Association journal (1990-07-01)
H K Xiao, S P Levine, G Kinnes, D Almaguer
PMID2382643
ABSTRACT
Results obtained using Fourier transform infrared spectrophotometry (FTIR) for the analysis of samples of carbon disulfide (CS2) eluates containing trichloroethylene (TCE) and Freon from charcoal air sampling tubes were evaluated by comparison with results obtained when using gas chromatography (GC). The FTIR yielded accurate results without regard to the presence of Freon.
Product Description
Supelco
1,1,2-Trichloro-1,2,2-trifluoroethane, certified reference material, 1000 μg/mL in methanol