Merck
  • Effect of Thunbergia laurifolia extract on extracellular dopamine level in rat nucleus accumbens.
Effect of Thunbergia laurifolia extract on extracellular dopamine level in rat nucleus accumbens.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-06-04)
Watchareewan Thongsaard, Charles Marsden
ABSTRACT
Thunbergia laurifolia Linn. (TL) is a herbal medicine used as an antidote for several poisonous agents in Thai traditional medicine. TL were reported not only to significantly increase potassium-stimulated dopamine release from rat striatal slices but also potentiated the effect of amphetamine on potassium-stimulated dopamine release. The present study aim to investigate the effect of TL on extracellular dopamine levels in rat nucleus accumbens in vivo in comparison to cocaine. Single injections of methanol extract of TL (200 mg/kg, ip) and cocaine (10 mg/kg, ip) were performed to determine the levels of extracellular dopamine in vivo in the living brain using the microdialysis technique (collecting time = 20 minutes). Cocaine and TL significantly (p < 0.05) increased extracellular dopamine levels in the rat nucleus accumbens in a similar manner, but the effect of cocaine had a faster onset than that of TL. The results are in agreement with a previous in vitro study. It can be summarized that TL stimulated rat dopamine release from the nucleus accumbens both in vitro and in vivo. This brain area is responsible for the reward mechanism of addiction. The clinical significance for the use of TL in humans and the effect of long term treatment with TL on addiction requires further investigation.
Sigma-Aldrich
Cocaine free base
Sigma-Aldrich
Cocaine hydrochloride
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis