Merck
  • Recyclable copper catalyzed nitrogenation of biphenyl halides: a direct approach to carbazoles.
Recyclable copper catalyzed nitrogenation of biphenyl halides: a direct approach to carbazoles.
Chemical communications (Cambridge, England) (2013-03-22)
Yang Ou, Ning Jiao
ABSTRACT
A novel A21-CuI catalyzed direct nitrogenation of biphenyl halides for the direct synthesis of carbazoles via a direct C-H amination process has been developed. A recyclable and inexpensive Cu-catalyst was successfully employed in N-heterocyclic compound synthesis via tandem azidation and C-H amination, which makes this protocol very practical and easy to handle.
Product Description
Supelco
Biphenyl solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Biphenyl, ≥99%
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%