Merck
  • Evaluation of transport parameters for PVC based polyvinyl alcohol Ce(IV) phosphate composite membrane.
Evaluation of transport parameters for PVC based polyvinyl alcohol Ce(IV) phosphate composite membrane.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Mohammad Mujahid Ali Khan, Rafiuddin, Inamuddin
ABSTRACT
The aim of this study was to investigate the preparation of novel membrane and the characterization of their properties. A new class of polyvinyl chloride (PVC) based polyvinyl alcohol Ce(IV) phosphate composite membrane was successfully prepared by solution casting method. The structural formation was confirmed by X-ray diffraction (XRD), Fourier transform infrared (FTIR) spectroscopy and morphological studies. The thermal property was investigated by thermogravimetry analysis (TGA) method. The order of surface charge density for various electrolytes was found to be LiCl<NaCl<KCl.
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~80,000, average Mn ~47,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed