Merck
  • Pressure-induced phase transition and polymerization of tetracyanoethylene (TCNE).
Pressure-induced phase transition and polymerization of tetracyanoethylene (TCNE).
The Journal of chemical physics (2013-03-15)
Dane Tomasino, Jing-Yin Chen, Minesob Kim, Choong-Shik Yoo
ABSTRACT
We have studied the pressure-induced physical and chemical transformations of tetracyanoethylene (TCNE or C6N4) in diamond anvil cells using micro-Raman spectroscopy, laser-heating, emission spectroscopy, and synchrotron x-ray diffraction. The results indicate that TCNE in a quasi-hydrostatic condition undergoes a shear-induced phase transition at 10 GPa and then a chemical change to two-dimensional (2D) C=N polymers above 14 GPa. These phase and chemical transformations depend strongly on the state of stress in the sample and occur sluggishly in non-hydrostatic conditions over a large pressure range between 7 and 14 GPa. The x-ray diffraction data indicate that the phase transition occurs isostructurally within the monoclinic structure (P21∕c) without any apparent volume discontinuity and the C=N polymer is highly disordered but remains stable to 60 GPa-the maximum pressure studied. On the other hand, laser-heating of the C=N polymer above 25 GPa further converts to a theoretically predicted 3D C-N network structure, evident from an emergence of new Raman νs(C-N) at 1404 cm(-1) at 25 GPa and the visual appearance of translucent solid. The C-N product is, however, unstable upon pressure unloading below 10 GPa, resulting in a grayish powder that can be considered as nano-diamonds with high-nitrogen content at ambient pressure. The C-N product shows a strong emission line centered at 640 nm at 30 GPa, which linearly shifts toward shorter wavelength at the rate of -1.38 nm∕GPa. We conjecture that the observed red shift upon unloading pressure is due to increase of defects in the C-N product and thereby weakening of C-N bonds.
Sigma-Aldrich
Tetracyanoethylene, 96%