Merck
Neonatal herpes simplex virus infections.
Pediatric clinics of North America (2013-03-14)
Swetha G Pinninti, David W Kimberlin
ABSTRACT
Neonatal herpes simplex virus infections are uncommon, but because of the morbidity and mortality associated with the infection they are often considered in the differential diagnosis of ill neonates. The use of polymerase chain reaction for diagnosis of central nervous system infections and the development of safe and effective antiviral therapy has revolutionized the diagnosis and management of these infants. Initiation of long-term antiviral suppressive therapy in these infants has led to significant improvement in morbidity. This article summarizes the epidemiology of neonatal herpes simplex virus infections and discusses clinical presentation, diagnosis, management, and follow up of infants with neonatal herpes disease.
Product Description
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Pricing and availability is not currently available.