- Site-specific conjugation of RAFT polymers to proteins via expressed protein ligation.
Site-specific conjugation of RAFT polymers to proteins via expressed protein ligation.
Chemical communications (Cambridge, England) (2013-02-21)
Yan Xia, Shengchang Tang, Bradley D Olsen
PMID23423478
ABSTRACT
Site-specific protein conjugates with RAFT polymers were synthesized using expressed protein ligation. Stable micelles were formed from both linear block copolymer and Y-shaped conjugates.