Merck
  • Synthesis and structural analysis of (imido)vanadium(V) complexes containing chelate (anilido)methyl-imine ligands: ligand effect in ethylene dimerization.
Synthesis and structural analysis of (imido)vanadium(V) complexes containing chelate (anilido)methyl-imine ligands: ligand effect in ethylene dimerization.
Inorganic chemistry (2013-02-20)
Kotohiro Nomura, Atsushi Igarashi, Shohei Katao, Wenjuan Zhang, Wen-Hua Sun
ABSTRACT
A series of (imido)vanadium dichlorido complexes containing chelate anionic donor ligands of the type, VCl2(L)(NR) [R = 1-adamantyl (Ad), L = 2-(2,6-Me2C6H3)NCH2(C9H6N) (2), 8-(2,6-Me2C6H3)NCH2(C9H6N) (3); L = 2-(2,6-R'2C6H3)NCH2(C5H4N), R = 2-MeC6H4, R' = Me (4a), (i)Pr (4b); L = 2-(2,6-Me2C6H3)NCH2(C5H4N), R = 4-MeC6H4 (5), 3,5-Me2C6H3 (6)], have been prepared and identified. The reactions with ethylene by 2,3 in the presence of methylaluminoxane (MAO) afforded a mixture of high molecular weight polyethylene and oligomers. Reactions with ethylene by VCl2[2-(2,6-R'2C6H3)NCH2(C5H4N)](NAd) (1a,b), 4-6 afforded 1-butene with high selectivities (>92%), and the activities by 4a,b are at the same level as those in 1a,b. The activities by 5,6 were lower than 4a,b and were at the same level of that by VCl2[2-(2,6-Me2C6H3)NCH2(C5H4N)](NPh). These results thus suggest that both the chelate anionic donor and the imido ligands play a role for both the activity and the selectivity.
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 1.0 mm, 99.7%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis