- Continuous synthesis and use of N-heterocyclic carbene copper(I) complexes from insoluble Cu2O.
Continuous synthesis and use of N-heterocyclic carbene copper(I) complexes from insoluble Cu2O.
Organic letters (2013-02-15)
Suzanne M Opalka, Jin Kyoon Park, Ashley R Longstreet, D Tyler McQuade
PMID23406503
ABSTRACT
It is demonstrated that homogeneous N-heterocyclic carbene-copper(I)-chloride complexes can be prepared continuously by flowing NHC precursors through a packed bed of solid Cu(2)O suspended in molecular sieves. The method enables the synthesis of a wide range of complexes including those that are challenging to prepare using standard approaches. Our strategy enables both sustained output of complex production for long-term catalytic reactions (greater than 5 h) and for generation of gram quantities for storage (greater than 1 g of complex in ~16 min).