Merck
  • Sequential analysis of masseter activity patterns during chewing in healthy males.
Sequential analysis of masseter activity patterns during chewing in healthy males.
Journal of medical engineering & technology (2013-01-31)
Yozo Miyaoka, Ichiro Ashida, Yuko Tamaki, Shin-ya Kawakami, Hajime Iwamori, Takako Yamazaki, Naoko Ito
ABSTRACT
The present study examined sequential changes in masseter activity patterns observed during chewing of four different agar samples in eight healthy young males. Two parameters, T(50) and D(50), were specifically used for evaluation of the activity patterns of individual bursts. Statistical significances were detected in regression coefficients (21.9% of 32 trials) and Spearman's rank correlation coefficients (28.1%) between the calculated T(50) values and chewing cycles, whereas no significant differences among the four agar samples were found. Three (I-III) types of activity patterns of masseter bursts during chewing sequences were classified by the D(50) values, which were derived from the T(50) values. The three types physiologically corresponded to incrementing (Type I), decrementing (Type III) and mixed discharge patterns (Type II). The classification of activity patterns suggested the usefulness of D(50) values in the sequential analysis of masseter activity patterns.
Product Description
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type A, ash 5-6%
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology