Merck
  • Novel regulation of cardiac metabolism and homeostasis by the adrenomedullin-receptor activity-modifying protein 2 system.
Novel regulation of cardiac metabolism and homeostasis by the adrenomedullin-receptor activity-modifying protein 2 system.
Hypertension (Dallas, Tex. : 1979) (2013-01-09)
Takahiro Yoshizawa, Takayuki Sakurai, Akiko Kamiyoshi, Yuka Ichikawa-Shindo, Hisaka Kawate, Yasuhiro Iesato, Teruhide Koyama, Ryuichi Uetake, Lei Yang, Akihiro Yamauchi, Megumu Tanaka, Yuichi Toriyama, Kyoko Igarashi, Tsutomu Nakada, Toshihide Kashihara, Mitsuhiko Yamada, Hayato Kawakami, Hiroki Nakanishi, Ryo Taguchi, Tsuyoshi Nakanishi, Hiroshi Akazawa, Takayuki Shindo
ABSTRACT
Adrenomedullin (AM) was identified as a vasodilating and hypotensive peptide mainly produced by the cardiovascular system. The AM receptor calcitonin receptor-like receptor associates with receptor activity-modifying protein (RAMP), one of the subtypes of regulatory proteins. Among knockout mice ((-/-)) of RAMPs, only RAMP2(-/-) is embryonically lethal with cardiovascular abnormalities that are the same as AM(-/-). This suggests that the AM-RAMP2 system is particularly important for the cardiovascular system. Although AM and RAMP2 are highly expressed in the heart from embryo to adulthood, their analysis has been limited by the embryonic lethality of AM(-/-) and RAMP2(-/-). For this study, we generated inducible cardiac myocyte-specific RAMP2(-/-) (C-RAMP2(-/-)). C-RAMP2(-/-) exhibited dilated cardiomyopathy-like heart failure with cardiac dilatation and myofibril disruption. C-RAMP2(-/-) hearts also showed changes in mitochondrial structure and downregulation of mitochondria-related genes involved in oxidative phosphorylation, β-oxidation, and reactive oxygen species regulation. Furthermore, the heart failure was preceded by changes in peroxisome proliferator-activated receptor-γ coactivator 1α (PGC-1α), a master regulator of mitochondrial biogenesis. Metabolome and matrix-assisted laser desorption/ionization-time-of-flight mass spectrometry (MALDI-TOF-MS) imaging analyses revealed early downregulation of cardiolipin, a mitochondrial membrane-specific lipid. Furthermore, primary-cultured cardiac myocytes from C-RAMP2(-/-) showed reduced mitochondrial membrane potential and enhanced reactive oxygen species production in a RAMP2 deletion-dependent manner. C-RAMP2(-/-) showed downregulated activation of cAMP response element binding protein (CREB), one of the main regulators of mitochondria-related genes. These data demonstrate that the AM-RAMP2 system is essential for cardiac metabolism and homeostasis. The AM-RAMP2 system is a promising therapeutic target of heart failure.
Sigma-Aldrich
Adrenomedullin 52 human, ≥90% (HPLC), powder
Sigma-Aldrich
Indium tin oxide coated glass slide, rectangular, surface resistivity 8-12 Ω/sq, slide