Merck
The tension of a curved surface from simulation.
The Journal of chemical physics (2012-12-27)
Alexander J Sodt, Richard W Pastor
ABSTRACT
This paper demonstrates a method for calculating the tension of a system with a curved interface from a molecular dynamics simulation. To do so, the pressure of a subset of the system is determined by applying a local (virtual) mechanical deformation, fitting the response to that of a bulk fluid, and then using the Young-Laplace equation to infer the tension of the interface. The accuracy of the method is tested by calculating the local pressure of a series of water simulations at various external pressures. The tension of a simulated curved octane-water interface is computed with the method and compares well with the planar tension (≈ 46.7 dyn/cm). Finally, an ambiguity is resolved between the Harasima and Irving-Kirkwood methods of calculating the local pressure as a means for computing the tension.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Octane, reagent grade, 98%