Merck
  • Comprehensive studies of the electronic structure of pristine and potassium doped chrysene investigated by electron energy-loss spectroscopy.
Comprehensive studies of the electronic structure of pristine and potassium doped chrysene investigated by electron energy-loss spectroscopy.
The Journal of chemical physics (2012-09-25)
Friedrich Roth, Benjamin Mahns, Ronny Schönfelder, Silke Hampel, Markus Nohr, Bernd Büchner, Martin Knupfer
ABSTRACT
We have performed electron energy-loss spectroscopy studies in order to investigate the electronic properties of chrysene molecular solids. The valence band electronic excitation spectra and the C 1s core level excitations have been measured for pristine and potassium doped chrysene. The core level studies show a fine structure which signals the presence of four close lying conduction bands close to the Fermi level. Upon potassium doping, these bands are filled with electrons, and we have reached a doping level of about K(2.7)chrysene. Furthermore, undoped chrysene is characterized by an optical gap of about 3.3 eV and five, relatively weak, excitonic features following the excitation onset. Doping induces major changes in the electronic excitation spectra, with a new, prominent low energy excitation at about 1.3 eV. The results of a Kramers-Kronig analysis indicate that this new feature can be assigned to a charge carrier plasmon in the doped material, and momentum dependent studies reveal a negative plasmon dispersion.
Product Description
Supelco
Chrysene solution, certified reference material, 1000 μg/mL in acetone
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland