Merck
  • Substituent-dependent fluorescent sensors for zinc ions based on carboxamidoquinoline.
Substituent-dependent fluorescent sensors for zinc ions based on carboxamidoquinoline.
Dalton transactions (Cambridge, England : 2003) (2012-08-21)
Yu Zhang, Xiangfeng Guo, Lihua Jia, Shicheng Xu, Zhihui Xu, Libo Zheng, Xuhong Qian
ABSTRACT
A series of carboxamidoquinoline-based fluorescent sensors (the AQZ family) were synthesized and characterized. The AQZ family members were highly soluble in water and showed good selectivity for Zn(2+)via enhanced fluorescence in aqueous buffer solution. Fluorescence signals could be tuned from dual-wavelength ratiometric changes to changes in the intensity of a single wavelength upon binding Zn(2+) through the introduction of different substituents onto the quinoline ring. Concentrations of free Zn(2+) of 10(-5)-10(-6) M could be detected using the sensors. Changes of substituents and their positions on the quinoline ring influenced the sensitivity for Zn(2+), but had little effect on Zn(2+) affinities.
Sigma-Aldrich
Quinoline, reagent grade, 98%
Sigma-Aldrich
Quinoline, reagent grade, 96%