Merck
  • Season's variation impact on Citrus aurantium leaves essential oil: chemical composition and biological activities.
Season's variation impact on Citrus aurantium leaves essential oil: chemical composition and biological activities.
Journal of food science (2012-08-18)
Inès Ellouze, Manef Abderrabba, Nassereddine Sabaou, Florence Mathieu, Ahmed Lebrihi, Jalloul Bouajila
ABSTRACT
Citrus aurantium leaves' essential oils (EOs) were evaluated for chemical composition and antioxidant and antibacterial activities. The vegetable material, taken 5 times during the year, has undergone the hydrodistillation to prepare EO. Chemical characterization by gas chromatography/mass spectrometry and GC/flame ionization detection allowed the identification of 46 compounds, and a notable quantitative and qualitative differences between the different Petitgrain samples according to the harvest time. Linalool (43.2% to 65.97%), linalyl acetate (0.77% to 24.77%), and α-terpineol (9.29% to 12.12%) were the main components. The most important number of components was registered for summer EOs (July and September). The 5 EOs submitted biological activities screening, namely, antioxidant and antimicrobial activities. Weak antioxidant activities (IC(50) values >10000 mg/L) were registered by both 1,1-diphenyl-2-picrylhydrazyl and 2,2'-azinobis-3-ethylbenzothiazoline-6-sulphonate assays, mostly because the weak amount of phenols in EOs. Antibacterial activities (12 microorganisms) were registered against Gram-positive bacteria [Bacillus subtilis (MIC = 2.7 mg/mL), Staphylococcus aureus (4.8 mg/mL)], and moderated ones against yeasts [Saccharomyces cerevisiae (9.2 mg/mL)] and fungi [Mucor ramannianus (5 mg/mL)]. Positive correlations between the identified compounds and the antimicrobial activities were noted. Many compounds were correlated to antimicrobial activity mainly caryophyllene oxide against Escherichia coli (R(2) = 0.99), S. cerevisiae (R(2) = 0.99), and Fusarium culmorum (R(2) = 0.99).
Product Description
Supelco
(+)-α-Terpineol, analytical standard
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Supelco
Linalool, analytical standard
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%