Merck
  • Cis-aconitic anhydride ethyl ester and phenolic compounds from the seeds of Alisma orientale.
Cis-aconitic anhydride ethyl ester and phenolic compounds from the seeds of Alisma orientale.
Natural product communications (2012-07-24)
Ming Zhao, Jing-Ying Chen, Li-Jia Xu, Tanja Goedecke, Xiao-Qi Zhang, Jin-Ao Duan, Chun-Tao Che
ABSTRACT
From the seeds of Alisma orientale, cis-aconitic anhydride ethyl ester and cis-2,4,5-trihydroxycinnamic acid were isolated, together with nine known phenolic compounds and a megastigmane sesquiterpene. All compounds are reported for the first time from Alisma species.
Sigma-Aldrich
cis-Aconitic anhydride, 95%