Merck
  • Cloning of a dibutyl phthalate hydrolase gene from Acinetobacter sp. strain M673 and functional analysis of its expression product in Escherichia coli.
Cloning of a dibutyl phthalate hydrolase gene from Acinetobacter sp. strain M673 and functional analysis of its expression product in Escherichia coli.
Applied microbiology and biotechnology (2012-06-26)
Jun Wu, Xuewei Liao, Fangbo Yu, Zhongbo Wei, Liuyan Yang
ABSTRACT
A dibutyl phthalate (DBP) transforming bacterium, strain M673, was isolated and identified as Acinetobacter sp. This strain could not grow on dialkyl phthalates, including dimethyl, diethyl, dipropyl, dibutyl, dipentyl, dihexyl, di(2-ethylhexyl), di-n-octyl, and dinonyl phthalate, but suspensions of cells could transform these compounds to phthalate via corresponding monoalkyl phthalates. During growth in Luria-Bertani medium, M673 produced the high amounts of non-DBP-induced intracellular hydrolase in the stationary phase. One DBP hydrolase gene containing an open reading frame of 1,095 bp was screened from a genomic library, and its expression product hydrolyzed various dialkyl phthalates to the corresponding monoalkyl phthalates.
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland