Merck
  • Selective production of 4-ethylphenolics from lignin via mild hydrogenolysis.
Selective production of 4-ethylphenolics from lignin via mild hydrogenolysis.
Bioresource technology (2012-06-22)
Yueyuan Ye, Yu Zhang, Juan Fan, Jie Chang
ABSTRACT
Selective production of 4-ethylphenolics from lignin via mild hydrogenolysis was reported in this short communication. The hydrogenolysis of lignin was carried out in an autoclave with 65 vol.% ethanol/water as solvent, with 5% Ru/C, Pd/C and Pt/C as catalysts. The influences of catalysts, lignin species, and reaction conditions including reaction temperature, reaction time, and initial H(2) pressure on yield of target compounds were investigated. 3.1% 4-Ethylphenol and 1.3% 4-ethylguaiacol based on lignin could be obtained simultaneously from hydrogenolysis of corn stalk lignin, which is approximate to the yield obtained from petrochemical route. The results of this work showed that this novel method is a quite promising technique for the substitution of petrochemical route.
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)