Merck
  • Argon-plasma-induced ultrathin thermal grafting of thermoresponsive pNIPAm coating for contractile patterned human SMC sheet engineering.
Argon-plasma-induced ultrathin thermal grafting of thermoresponsive pNIPAm coating for contractile patterned human SMC sheet engineering.
Macromolecular bioscience (2012-04-27)
Shahrzad Rayatpisheh, Peng Li, Mary B Chan-Park
ABSTRACT
A new method for ultrathin grafting of pNIPAm on PDMS surfaces is introduced that employs plasma activation of the surface followed by thermal polymerization. This method is optimized for human primary SMC attachment and subsequent intact cell sheet detachment by lowering the temperature. The contractile gene expression of the cells showed that the contractile phenotype of the SMCs which is induced by aligning the cells through micropatterning is more preserved after thermoresponsive cell sheet detachment in contrast with enzymatic detachment. Given its simplicity and low cost, this thermoresponsive grafting method can be utilized for engineering patterned cell sheets for future bottom-up tissue engineering techniques.
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Pricing and availability is not currently available.