Merck
  • Dihydroceramide accumulation and reactive oxygen species are distinct and nonessential events in 4-HPR-mediated leukemia cell death.
Dihydroceramide accumulation and reactive oxygen species are distinct and nonessential events in 4-HPR-mediated leukemia cell death.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2012-03-21)
Aintzane Apraiz, Jolanta Idkowiak-Baldys, Naiara Nieto-Rementería, María Dolores Boyano, Yusuf A Hannun, Aintzane Asumendi
ABSTRACT
4-(Hydroxyphenyl)retinamide (4-HPR) is a synthetic retinoid with a strong apoptotic effect towards different cancer cell lines in vitro, and it is currently tested in clinical trials. Increases of reactive oxygen species (ROS) and modulation of endogenous sphingolipid levels are well-described events observed upon 4-HPR treatment, but there is still a lack of understanding of their relationship and their contribution to cell death. LC-MS analysis of sphingolipids revealed that in human leukemia CCRF-CEM and Jurkat cells, 4-HPR induced dihydroceramide but not ceramide accumulation even at sublethal concentrations. Myriocin prevented the 4-HPR-induced dihydroceramide accumulation, but it did not prevent the loss of viability and increase of intracellular ROS production. On the other hand, ascorbic acid, Trolox, and vitamin E reversed 4-HPR effects on cell death but not dihydroceramide accumulation. NDGA, described as a lipoxygenase inhibitor, exerted a significantly higher antioxidant activity than vitamin E and abrogated 4-HPR-mediated ROS. It did not however rescue cellular viability. Taken together, this study demonstrates that early changes observed upon 4-HPR treatment, i.e., sphingolipid modulation and ROS production, are mechanistically independent events. Furthermore, the results indicate that 4-HPR-driven cell death may occur even in the absence of dihydroceramide or ROS accumulation. These observations should be taken into account for an improved design of drug combinations.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥90% (HPLC), from Larrea divaricata (creosote bush)
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
Retinoic acid p-hydroxyanilide, ≥95%