Merck
  • Quality control of modified xiaoyao san through the determination of 22 active components by ultra-performance liquid chromatography.
Quality control of modified xiaoyao san through the determination of 22 active components by ultra-performance liquid chromatography.
Journal of AOAC International (2012-02-11)
Feng Qin, Jun Huang, Xinjian Qiu, Sihang Hu, Xi Huang
ABSTRACT
A simple, sensitive, and reliable ultra-performance liquid chromatography (UPLC) method has been developed for simultaneous determination of 22 major constituents in modified xiaoyao san (MXS), a multiherbal formula. The chromatographic separation was performed on an ACQUITY UPLC BEH C18 column (150 x 2.1 mm, 1.7 microm, particle size), with an aqueous 0.5% acetic acid and acetonitrile mobile phase gradient. The method was validated for linearity (r2 >0.9937), intraday and interday precision (RSD <8.51%), recovery (91.18-107.73%), LOD (0.02-4.17 ng/mL), and LOQ (0.05-12.50 ng/mL). The established method was successfully applied to quantify the 22 marker compounds in MXS, which provided a useful basis of overall evaluation of the quality of MXS.
Sigma-Aldrich
2′-Hydroxy-4′-methoxyacetophenone, 99%