Merck
Colpodella spp.-like parasite infection in woman, China.
Emerging infectious diseases (2012-01-21)
Cong L Yuan, Patrick J Keeling, Peter J Krause, Ales Horak, Stephen Bent, Lindsay Rollend, Xiu G Hua
ABSTRACT
The phylum Apicomplexa comprises intracellular protozoa that include many human pathogens. Their nearest relatives are chromerids and colpodellids. We report a case of a Babesia spp.-like relapsing infection caused by a newly described microorganism related to the Apicomplexa. This case is highly suggestive of a previously undescribed type of colpodellid that infects vertebrates.
Sigma-Aldrich
Artemether, ≥98% (HPLC)
Pricing and availability is not currently available.