Merck
  • Application of density functional theory (DFT) to study the properties and degradation of natural estrogen hormones with chemical oxidizers.
Application of density functional theory (DFT) to study the properties and degradation of natural estrogen hormones with chemical oxidizers.
The Science of the total environment (2012-01-17)
Ekaterina V Rokhina, Rominder P S Suri
ABSTRACT
Estrone (E1), 17β-estradiol (E2), estriol (E3), equilin (EQ) and 17α-estradiol (17α) estrogen hormones are released by humans and animals and have been detected in the environment and municipal wastewater treatment plants. The structural and electronic properties of natural hormone molecules are investigated by performing density functional theory calculations and used to predict their properties and chemical behavior. Quantitative structure property relationship (QSPR) approach is applied to correlate the estrogenicity associated with the natural estrogen hormones according to their molecular properties. The obtained relationship reveals the importance of the frontier molecular orbital energy in the interpretation of estrogenic activity of hormones, which is consistent with the previous research. Moreover, the obtained molecular descriptors also aid determination of the degradability of hormones, and to rationalize degradation pathways, with chemical oxidizers such as ozone and hydroxyl radical. Both types of interactions belong to the orbital-controlled reactions. The active sites determined by Fukui functions for the estrogen hormone molecules confirm the reaction pattern that initiates the attack of the aromatic ring for both ozone and hydroxyl radical. The reactive sites of the molecules are mapped with subsequent reaction intermediates and compared with experimental data obtained from the literature.
Product Description
Supelco
Equilin, VETRANAL®, analytical standard
Sigma-Aldrich
Equilin