Merck
  • ortho-Lithium/magnesium carboxylate-driven aromatic nucleophilic substitution reactions on unprotected naphthoic acids.
ortho-Lithium/magnesium carboxylate-driven aromatic nucleophilic substitution reactions on unprotected naphthoic acids.
The Journal of organic chemistry (2011-11-24)
Regadia Aissaoui, Arnaud Nourry, Ariane Coquel, Thi Thanh Hà Dao, Aicha Derdour, Jean-Jacques Helesbeux, Olivier Duval, Anne-Sophie Castanet, Jacques Mortier
ABSTRACT
Substitution of an ortho-fluoro or methoxy group in 1- and 2-naphthoic acids furnishing substituted naphthoic acids occurs in good to excellent yields upon reaction with alkyl/vinyl/aryl organolithium and Grignard reagents, in the absence of a metal catalyst without the need to protect the carboxyl (CO(2)H) group. This novel nucleophilic aromatic substitution is presumed to proceed via a precoordination of the organometallic with the substrate, followed by an addition/elimination.
Sigma-Aldrich
2-Naphthoic acid, 98%
Sigma-Aldrich
1-Naphthoic acid, 96%