Merck
Membrane labeling and immobilization via copper-free click chemistry.
Chemical communications (Cambridge, England) (2011-10-04)
Heidi E Bostic, Matthew D Smith, Andrei A Poloukhtine, Vladimir V Popik, Michael D Best
ABSTRACT
Copper-free click chemistry was employed to derivatize membrane bilayers. This approach uses an azido-lipid conjugate presented on liposomes, which can be labeled in bioorthogonal fashion via cyclooctyne-tagged reagents. An immobilization-based approach using streptavidin-coated microplates was exploited to evaluate membrane derivatization.
Sigma-Aldrich
Cyclooctane, ≥99%