Merck
  • Study of influential factors on oligosaccharide formation by fructosyltransferase activity during stachyose hydrolysis by Pectinex ultra SP-L.
Study of influential factors on oligosaccharide formation by fructosyltransferase activity during stachyose hydrolysis by Pectinex ultra SP-L.
Journal of agricultural and food chemistry (2011-09-03)
Antonia Montilla, Agustín Olano, Cristina Martínez-Villaluenga, Nieves Corzo
ABSTRACT
The influence of reaction conditions for oligosaccharide synthesis from stachyose using a commercial enzymatic preparation from Aspergillus aculeatus (Pectinex Ultra SP-L) was studied. Oligosaccharides were analyzed by gas chromatography with flame ionization detection (GC-FID) and matrix-assisted laser desorption/ionization-time-of-flight-mass spectrometry (MALDI-TOF-MS). Galactosyl-melibiose (DP(3)) was synthesized as a result of fructosidase activity, whereas fructosyl-stachyose (DP(5)) and difructosyl-stachyose (DP(6)) were formed as a consequence of the fructosyltransferase activity of Pectinex Ultra SP-L. The optimal reaction conditions for the synthesis of penta- and hexasaccharides were 60 °C, pH 5.5, 600 mg/mL stachyose, and 34 U/mL enzyme. Reaction time played an important role in oligosaccharide mixture composition constituted by 20% DP(5), 0.7% DP(6), 55% stachyose, 21% galactosyl-melibiose, and 1% monosaccharides after 1 h and 16% DP(5), 4% DP(6), 27% stachyose, 44% galactosyl-melibiose, and 2% monosaccharides after 3 h. In conclusion, stachyose could be used as a substrate for the enzymatic synthesis of new oligosaccharides that may open new opportunities in the development of future prebiotics.
Sigma-Aldrich
Stachyose hydrate, ≥98% (HPLC)