Merck
  • A comparison of Tris-glycine and Tris-tricine buffers for the electrophoretic separation of major serum proteins.
A comparison of Tris-glycine and Tris-tricine buffers for the electrophoretic separation of major serum proteins.
Journal of separation science (2011-08-06)
Syed R Haider, Barry L Sharp, Helen J Reid
ABSTRACT
This paper compares different buffer systems for the electrophoretic separation of the five most abundant serum proteins on native-PAGE gel and cellulose membranes. A modified Tris-tricine system was shown to be superior for the separation of these serum proteins in a 7% m/v native-PAGE gel as compared with the traditionally used Tris-glycine and Tris-tricine methods. This modified Tris-tricine buffer system was also employed for the separation of serum proteins using a cellulose acetate membrane and very effective separation was observed as compared with the traditionally used Tris-barbital and Tris-glycine buffer systems.
Product Description
SAFC
Tricine
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)