Merck
  • A catabolic pathway for the degradation of chrysene by Pseudoxanthomonas sp. PNK-04.
A catabolic pathway for the degradation of chrysene by Pseudoxanthomonas sp. PNK-04.
FEMS microbiology letters (2011-05-07)
Anand S Nayak, Sanganal Sanjeev Kumar, Mudde Santosh Kumar, Oblesha Anjaneya, Timmanagouda B Karegoudar
ABSTRACT
The chrysene-degrading bacterium Pseudoxanthomonas sp. PNK-04 was isolated from a coal sample. Three novel metabolites, hydroxyphenanthroic acid, 1-hydroxy-2-naphthoic acid and salicylic acid, were identified by TLC, HPLC and MS. Key enzyme activities, namely 1-hydroxy-2-naphthoate hydroxylase, 1,2-dihydroxynaphthalene dioxygenase, salicylaldehyde dehydrogenase and catechol-1,2-dioxygenase, were noted in the cell-free extract. These results suggest that chrysene is catabolized via hydroxyphenanthroic acid, 1-hydroxy-2-naphthoic acid, salicylic acid and catechol. The terminal aromatic metabolite, catechol, is then catabolized by catechol-1,2-dioxygenase to cis,cis-muconic acid, ultimately forming TCA cycle intermediates. Based on these studies, the proposed catabolic pathway for chrysene degradation by strain PNK-04 is chrysene → hydroxyphenanthroic acid → 1-hydroxy-2-naphthoic acid → 1,2-dihydroxynaphthalene → salicylic acid → catechol →cis,cis-muconic acid.
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Supelco
Chrysene solution, certified reference material, 1000 μg/mL in acetone
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material
Sigma-Aldrich
1-Naphthoic acid, 96%
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland