Merck
  • Molecular aspects of glucose dehydration by chromium chlorides in ionic liquids.
Molecular aspects of glucose dehydration by chromium chlorides in ionic liquids.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-04-14)
Yanmei Zhang, Evgeny A Pidko, Emiel J M Hensen
ABSTRACT
A combined experimental and computational study of the ionic-liquid-mediated dehydration of glucose and fructose by Cr(II) and Cr(III) chlorides has been performed. The ability of chromium to selectively dehydrate glucose to 5-hydroxymethylfurfural (HMF) in the ionic liquid 1-ethyl-3-methyl imidazolium chloride does not depend on the oxidation state of chromium. Nevertheless, Cr(III) exhibits higher activity and selectivity to HMF than Cr(II) . Anhydrous CrCl(2) and CrCl(3)⋅6 H(2)O readily catalyze glucose dehydration with HMF yields of 60 and 72%, respectively, after 3 h. Anhydrous CrCl(3) has a lower activity, because it only slowly dissolves in the reaction mixture. The transformation of glucose to HMF involves the formation of fructose as an intermediate. The exceptional catalytic performance of the chromium catalysts is explained by their unique ability to catalyze glucose to fructose isomerization and fructose to HMF dehydration with high selectivity. Side reactions leading to humins by means of condensation reactions take predominantly place during fructose dehydration. The higher HMF selectivity for Cr(III) is tentatively explained by the higher activity in fructose dehydration compared to Cr(II) . This limits the concentration of intermediates that are involved in bimolecular condensation reactions. Model DFT calculations indicate a substantially lower activation barrier for glucose isomerization by Cr(III) compared to Cr(II) . Qualitatively, glucose isomerization follows a similar mechanism for Cr(II) and Cr(III) . The mechanism involves ring opening of D-glucopyranose coordinated to a single Cr ion, followed by a transient self-organization of catalytic chromium complexes that promotes the rate-determining hydrogen-shift step.
Sigma-Aldrich
Chromium(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Chromium(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Chromium(II) chloride, 95%