Merck
  • A novel and comprehensive mouse model of human non-alcoholic steatohepatitis with the full range of dysmetabolic and histological abnormalities induced by gold thioglucose and a high-fat diet.
A novel and comprehensive mouse model of human non-alcoholic steatohepatitis with the full range of dysmetabolic and histological abnormalities induced by gold thioglucose and a high-fat diet.
Liver international : official journal of the International Association for the Study of the Liver (2011-03-09)
Mitsunari Ogasawara, Akira Hirose, Masafumi Ono, Kosuke Aritake, Yasuko Nozaki, Masaya Takahashi, Nobuto Okamoto, Shuji Sakamoto, Shinji Iwasaki, Taketoshi Asanuma, Taketoshi Taniguchi, Yoshihiro Urade, Saburo Onishi, Toshiji Saibara, Jude A Oben
ABSTRACT
The search for effective treatments of non-alcoholic steatohepatitis (NASH), now the most common chronic liver disease in affluent countries, is hindered by a lack of animal models having the range of anthropometric and pathophysiological features as human NASH. To examine if mice treated with gold thioglucose (GTG) - known to induce lesions in the ventromedial hypothalamus, leading to hyperphagia and obesity - and then fed a high-fat diet (HF) had a comprehensive histological and dysmetabolic phenotype resembling human NASH. C57BL/6 mice were injected intraperitoneally with GTG and then fed HF for 12 weeks (GTG+HF). The extent of abdominal adiposity was assayed by CT scanning. A glucose tolerance test and an insulin tolerance test were performed to evaluate insulin resistance (IR). Histological, molecular and biochemical analyses were also performed. Gold thioglucose+HF induced dysmetabolism, with hyperphagia, obesity with increased abdominal adiposity, IR and consequent steatohepatitis, with hepatocyte ballooning, Mallory-Denk bodies, perivenular and pericellular fibrosis as seen in adult NASH, paralleled by an increased expression of the profibrogenic factors, transforming growth factor-β1 and TIMP-1. Plasma adiponectin and the expression of adiponectin receptor 1 and receptor 2 were decreased, while PPAR-γ and FAS were increased in the livers of GTG+HF mice. In addition, GTG+HF mice showed glucose intolerance and severe IR. Treatment with GTG and HF diet induce, in mice, a comprehensive model of human NASH, with the full range of dysmetabolic and histological abnormalities.
Sigma-Aldrich
Aurothioglucose hydrate, ≥96% (titration)