Merck
  • C2-symmetric chiral disulfoxide ligands in rhodium-catalyzed 1,4-addition: from ligand synthesis to the enantioselection pathway.
C2-symmetric chiral disulfoxide ligands in rhodium-catalyzed 1,4-addition: from ligand synthesis to the enantioselection pathway.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-11-16)
Ronaldo Mariz, Albert Poater, Michele Gatti, Emma Drinkel, Justus J Bürgi, Xinjun Luan, Sascha Blumentritt, Anthony Linden, Luigi Cavallo, Reto Dorta
ABSTRACT
A family of chiral C(2)-symmetric disulfoxide ligands possessing biaryl atropisomeric backbones has been synthesized by using the Andersen methodology. Complete characterization includes X-ray crystallographic studies of all ligands and some of their rhodium complexes. Their synthesis, optical purity, electronic properties, and catalytic behavior in the prototypical rhodium-catalyzed 1,4-addition of phenylboronic acid to 2-cyclohexen-1-one are presented through an in depth study of this ligand class. Density functional theory calculations on the step of the catalytic cycle that determines the enantioselectivity are presented and reinforce the first hypothetical explanations for the high levels of asymmetric induction observed.
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%
Pricing and availability is not currently available.