- Kinetics of sorption-desorption of benfuracarb insecticide in mollisols.
Kinetics of sorption-desorption of benfuracarb insecticide in mollisols.
Sorption-desorption processes govern the movement of pesticides in soil. These processes determine the potential hazard of the pesticide in a given environment for groundwater contamination and need to be investigated. In the present study, sorption-desorption processes of benfuracarb were investigated using a batch method in two mollisols. The kinetics of benfuracarb sorption in mollisols conformed to two-compartment (1 + 1) first-order kinetics. The fast sorption rate constant was about 3 times higher for silt loam than for loam soil. However, the slow sorption rate constants were statistically similar for both soils. The concentration-dependent sorption-desorption isotherms of benfuracarb could not closely conform to the Freundlich isotherm in mollisols of high organic C content. The computed values of both the sorption (log K) and desorption (log K') capacities were higher for silt loam than for loam soil. The desorption index (n'/n) values in the range 30.0-41.3 indicated poor reversibility of sorbed benfuracarb in mollisols. In view of the strong sorption of benfuracarb in mollisols with only partial desorption, the possibility of the leaching of soil-applied benfuracarb to contaminate groundwaters appears to be low.