Merck
  • Pharmacological inhibition of intracellular calcium release blocks acid-induced bone resorption.
Pharmacological inhibition of intracellular calcium release blocks acid-induced bone resorption.
American journal of physiology. Renal physiology (2010-11-05)
Nancy S Krieger, David A Bushinsky
ABSTRACT
In vivo chronic metabolic acidosis induces net Ca2+ efflux from bone, and incubation of neonatal mouse calvariae in medium simulating physiological metabolic acidosis induces bone resorption. It appears that activation of the proton (H+) receptor OGR1 in the osteoblast leads to an increase in intracellular Ca2+, which is associated with an increase in cyclooxygenase 2 (COX2) and PGE2-induced receptor activator of NF-κB ligand (RANKL) and H+-induced osteoclastic bone resorption. To support this hypothesis, we tested whether intracellular Ca2+ signaling was integral to H+-induced bone resorption by determining whether 8-(N,N-diethylamino)octyl-3,4,5-trimethoxybenzoate (TMB-8) and 2-aminoethoxydiphenyl borate (2-APB), inhibitors of inositol trisphosphate-mediated Ca2+ signaling, would block H+-induced bone resorption in cultured neonatal calvariae and, if so, would do so by inhibiting H+-induced stimulation of COX2 and RANKL in osteoblastic cells. We found that H+-induced bone resorption is significantly inhibited by TMB-8 and 2-APB. Both compounds also inhibit H+-induced stimulation of COX2 protein in calvariae and COX2 mRNA and protein levels in primary osteoblasts. H+-induced stimulation of RANKL in calvarial cultures, as well as primary cells, is also completely inhibited by TMB-8 and 2-APB. These results support the hypothesis that H+ stimulation of net Ca2+ efflux from bone, mediated by COX2- and subsequent PGE2-induced RANKL production, is initiated in the osteoblast via activation of Ca2+ signaling.
Sigma-Aldrich
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride, ≥98%