Merck
Metabolism and toxicity of menthofuran in rat liver slices and in rats.
Chemical research in toxicology (2010-10-16)
S Cyrus Khojasteh, Shimako Oishi, Sidney D Nelson
ABSTRACT
Menthofuran is a monoterpene present in mint plants that is oxidized by mammalian cytochrome P450 (CYP) to hepatotoxic metabolites. Evidence has been presented that p-cresol and other unusual oxidative products are metabolites of menthofuran in rats and that p-cresol may be responsible in part for the hepatotoxicity caused by menthofuran [ Madyastha, K. M. and Raj, C. P. (1992) Drug Metab. Dispos. 20, 295 - 301]. In the present study, several oxidative metabolites of menthofuran were characterized in rat and human liver microsomes and in rat liver slices exposed to cytotoxic concentrations of menthofuran. Metabolites that were identified were monohydroxylation products of the furanyl and cyclohexyl groups, mintlactones and hydroxymintlactones, a reactive γ-ketoenal, and a glutathione conjugate. A similar spectrum of metabolites was found in urine 24 h after the administration of hepatotoxic doses of menthofuran to rats. In no case was p-cresol (or any of the other reported unusual oxidative metabolites of menthofuran) detected above background concentrations that were well below concentrations of p-cresol that cause cytotoxicity in rat liver slices. Thus, the major metabolites responsible for the hepatotoxic effects of menthofuran appear to be a γ-ketoenal and/or epoxides formed by oxidation of the furan ring.
Product Description
Supelco
(+)-Menthofuran, analytical standard
Sigma-Aldrich
4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran, ≥95%