Merck
  • [Preparation of magnetic latexes and their use for the immunodetection of microbial antigens].
[Preparation of magnetic latexes and their use for the immunodetection of microbial antigens].
Prikladnaia biokhimiia i mikrobiologiia (2010-09-29)
S M Kal'noĭ, I V Zharnikova, A A Zaĭtsev, A I Bondarenko, I Iu Borzdova, V V Ostapovich, A A Kurilova
ABSTRACT
The possibility of detecting antigens of plague, tularemia, and brucellosis microbes with magnetic latex (ML)-based test systems has been demonstrated. MLs were prepared from latexes (polyacroleine microspheres, 1.2-1.8 +/- 0.1 microm) by exposing the particles to a 25-35%-solution of ferrous sulfate for 0.5 h and then to a 15-25%-aqueous solution of ammonia for 0.5 h in a 100 degrees C water bath and dehydrating after each operation. The possibility of preparing magnetic latex immunosorbents (MLIS) by ligand immobilization on ML and using them in magnetic latex ELISA (ML-ELISA) for the detection of microbial antigens was demonstrated. The detection limit in ML-ELISA equaled 10(2)-10(3) microbial cells in 1 ml (cells/ml). Relative experimental error was not higher than 8%.
Sigma-Aldrich
Iron(II) oxide, −10 mesh, ≥99.6% trace metals basis