Merck
Volatile constituents of two rare subspecies of Thymus praecox.
Natural product communications (2010-08-26)
Danijela Vidic, Sanja Cavar, Marija Edita Solić, Milka Maksimović
ABSTRACT
Hydrodistilled essential oil and the corresponding headspace volatiles of Bosnian wild growing Thymus praecox ssp. polytrichus (A. Kern. Ex Borbàs) Jalas and Thymus praecox ssp. skorpilii (Velen.) Jalas were subjected to capillary GC-MS analysis. This work presents a detailed essential oil analysis of these two rare Thymus subspecies from Bosnia, as well as the very first report on their headspace composition. Eighty-seven volatile constituents were identified in four samples. Two alcohols were the major constituents in the essential oil of T. praecox ssp. polytrichus, a monoterpene, linalool (13.9%), and a sesquiterpene, (E)-nerolidol (10.4%), while linalyl acetate (36.7%) and linalool (22.7%) were the most abundant volatiles in the corresponding headspace sample. Oxygenated monoterpenes (57.5%) predominate in the essential oil of T. praecox ssp. skorpilii with linalyl acetate (28.7%) and linalool (14.4%) as the main representatives. Headspace sample of this subspecies also showed richness in linalyl acetate (52.4%), while the second most abundant compound was alpha-pinene (14.5%), a monoterpene hydrocarbon.
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%