Merck
  • Highly efficient procedure for the synthesis of fructone fragrance using a novel carbon based acid.
Highly efficient procedure for the synthesis of fructone fragrance using a novel carbon based acid.
Molecules (Basel, Switzerland) (2010-08-18)
Baowei Hu, Chunqing Li, Sheng-Xian Zhao, Lin-Mei Rong, Shao-Qin Lv, Xuezheng Liang, Chenze Qi
ABSTRACT
The novel carbon based acid has been synthesized via one-step hydrothermal carbonization of furaldehyde and hydroxyethylsulfonic acid. A highly efficient procedure for the synthesis of fructone has been developed using the novel carbon based acid. The results showed that the catalyst possessed high activity for the reaction, giving a yield of over 95%. The advantages of high activity, stability, reusability and low cost for a simple synthesis procedure and wide applicability to various diols and beta-keto esters make this novel carbon based acid one of the best choices for the reaction.
Sigma-Aldrich
Isethionic acid ammonium salt, 99%
Sigma-Aldrich
Isethionic acid sodium salt, 98%