Merck
  • UV-induced tolerance to a contact allergen is impaired in polymorphic light eruption.
UV-induced tolerance to a contact allergen is impaired in polymorphic light eruption.
The Journal of investigative dermatology (2010-07-09)
Leena M Koulu, Jarmo K Laihia, Hanna-Helena Peltoniemi, Christer T Jansén
ABSTRACT
Polymorphic light eruption (PLE) is a common skin disorder provoked by exposure to UVR. Its clinical symptoms resemble those of a contact allergic reaction. PLE is generally considered a T-cell-mediated autoimmune reaction toward a yet unidentified antigen formed in UVR-exposed skin. Predisposition to such an immune reaction may result from aberrant epitope formation, increased immune reactivity to a universal epitope, or diminished propensity to UVR-induced immunosuppression or to the induction of tolerance. In a study comprising a total of 24 PLE patients and 24 healthy sex- and age-matched controls, we found that both groups demonstrated similar immunosuppression of contact sensitization to diphenylcyclopropenone by earlier exposure to solar-simulating UVR. However, only 1 out of 13 PLE patients (8%) versus 6 out of 11 controls (55%) that had been immunosuppressed by UVR exhibited a state of immunotolerance toward the same allergen after 10-24 months (P=0.023). We conclude that the impaired propensity to UVR-induced allergen-specific immunotolerance may promote recurrent PLE.
Sigma-Aldrich
Diphenylcyclopropenone, 98%