Merck
  • Transient linear eruption: asymmetric periflexural exanthem or blaschkitis.
Transient linear eruption: asymmetric periflexural exanthem or blaschkitis.
Pediatric dermatology (2010-07-09)
Bavani Arun, Asad Salim
ABSTRACT
We report the case of a 4-year-old boy who presented with a transient extensive eczematous eruption on the left side of his trunk and extremities, with clear demarcation in the midline. The eruption started to resolve without any sequelae in 4 weeks. The history and clinical findings suggest that this transient eruption could have been either a case of unilateral blaschkitis in childhood or asymmetric periflexural exanthem.
Product Description
Clobetasone butyrate for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Clobetasone butyrate, ≥98%
Pricing and availability is not currently available.
Clobetasone butyrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.