Merck
  • Strained to the limit: when a cyclobutyl moiety becomes a thermodynamic sink in a protolytic ring-opening of photogenerated oxetanes.
Strained to the limit: when a cyclobutyl moiety becomes a thermodynamic sink in a protolytic ring-opening of photogenerated oxetanes.
Organic letters (2010-07-09)
Roman A Valiulin, Teresa M Arisco, Andrei G Kutateladze
ABSTRACT
Strained polycyclic oxetanes generated photochemically from the Diels-Alder adducts of cyclic dienes and enones undergo deep skeletal rearrangements under protolytic ring-opening conditions offering expeditious access to chlorohydrins and other products of unique skeletal topology.
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%
Sigma-Aldrich
2-Cyclohexen-1-one, produced by BASF, ≥98.0% (GC)