Merck
  • Genotoxicity study of photolytically treated 2-chloropyridine aqueous solutions.
Genotoxicity study of photolytically treated 2-chloropyridine aqueous solutions.
Journal of hazardous materials (2010-01-20)
Dimitris Vlastos, Charalambos G Skoutelis, Ioannis T Theodoridis, David R Stapleton, Maria I Papadaki
ABSTRACT
2-Chloropyridine (2-CPY) has been identified as a trace organic chemical in process streams, wastewater and even drinking water. Furthermore, it appears to be formed as a secondary pollutant during the decomposition of specific insecticides. As reported in our previous work, 2-CPY was readily removed and slowly mineralised when subjected to ultraviolet (UV) irradiation at 254 nm. Moreover, 2-CPY was found to be genotoxic at 100 microg ml(-1) but it was not genotoxic at or below 50 microg ml(-1). In this work 2-CPY aqueous solutions were treated by means of UV irradiation at 254 nm. 2-CPY mineralisation history under different conditions is shown. 2-CPY was found to mineralise completely upon prolonged irradiation. Identified products of 2-CPY photolytic decomposition are presented. Solution genotoxicity was tested as a function of treatment time. Aqueous solution samples, taken at different photo-treatment times were tested in cultured human lymphocytes applying the cytokinesis block micronucleus (CBMN) assay. It was found that the solution was genotoxic even when 2-CPY had been practically removed. This shows that photo-treatment of 2-CPY produces genotoxic products. Upon prolonged irradiation solution genotoxicity values approached the control value.
Sigma-Aldrich
2-Chloropyridine, 99%