Merck
  • Structure and inhibition of the SARS coronavirus envelope protein ion channel.
Structure and inhibition of the SARS coronavirus envelope protein ion channel.
PLoS pathogens (2009-07-14)
Konstantin Pervushin, Edward Tan, Krupakar Parthasarathy, Xin Lin, Feng Li Jiang, Dejie Yu, Ardcharaporn Vararattanavech, Tuck Wah Soong, Ding Xiang Liu, Jaume Torres
ABSTRACT
The envelope (E) protein from coronaviruses is a small polypeptide that contains at least one alpha-helical transmembrane domain. Absence, or inactivation, of E protein results in attenuated viruses, due to alterations in either virion morphology or tropism. Apart from its morphogenetic properties, protein E has been reported to have membrane permeabilizing activity. Further, the drug hexamethylene amiloride (HMA), but not amiloride, inhibited in vitro ion channel activity of some synthetic coronavirus E proteins, and also viral replication. We have previously shown for the coronavirus species responsible for severe acute respiratory syndrome (SARS-CoV) that the transmembrane domain of E protein (ETM) forms pentameric alpha-helical bundles that are likely responsible for the observed channel activity. Herein, using solution NMR in dodecylphosphatidylcholine micelles and energy minimization, we have obtained a model of this channel which features regular alpha-helices that form a pentameric left-handed parallel bundle. The drug HMA was found to bind inside the lumen of the channel, at both the C-terminal and the N-terminal openings, and, in contrast to amiloride, induced additional chemical shifts in ETM. Full length SARS-CoV E displayed channel activity when transiently expressed in human embryonic kidney 293 (HEK-293) cells in a whole-cell patch clamp set-up. This activity was significantly reduced by hexamethylene amiloride (HMA), but not by amiloride. The channel structure presented herein provides a possible rationale for inhibition, and a platform for future structure-based drug design of this potential pharmacological target.
Sigma-Aldrich
5-(N,N-Hexamethylene)amiloride
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder